Learn More
Electron Microscopy Sciences Frozen Section Brush, 4 per set
SDP
Catalog No. 50364874
Encompass_Preferred
These flat white boar bristle brush which is cut into an angle. This stiffness bristle will prevent the frozen section from curling through the brush. We offer a set of four brushes: two #1 with 3/8" angle cut and #2 with 1/4" angle cut.
Catalog No. 50-364-874 Supplier Electron Microscopy Sciences Supplier No. 6173501
These flat white boar bristle brush which is cut into an angle. This stiffness bristle will prevent the frozen section from curling through the brush. We offer a set of four brushes: two #1 with 3/8" angle cut and #2 with 1/4" angle cut.