Learn More
Biochain Institute Inc Frozen Tissue Array - 24 Different Prostate Tumors and 4 Corresponding Normal Controls, 1 slide/PK
SDP
Catalog No. 50180884
Encompass_Preferred
BioChain's tissue products are based on the sample repository network established following the IRB-approved ethical standard and procedures. BioChain's human tumor frozen tissue arrays are designed for studying tumor tissue specific cellular localization of genes or proteins of interest in different human tissues. Same kind of tumors and corresponding normal tissues are mounted on positively charged slides.
Catalog No. 50-180-884 Supplier Biochain Institute Inc Supplier No. T6235201
Add to Cart
Add to Cart
BioChain's tissue products are based on the sample repository network established following the IRB-approved ethical standard and procedures. BioChain's human tumor frozen tissue arrays are designed for studying tumor tissue specific cellular localization of genes or proteins of interest in different human tissues. Same kind of tumors and corresponding normal tissues are mounted on positively charged slides.