Learn More
Biochain Institute Inc Frozen Tissue Array - Rabbit Female Adult Normal Tissue, Multi-tissue I, 5 slides/PK
SDP
Supplier:  Biochain Institute Inc T6834700G025
Encompass_Preferred
BioChain's rabbit normal multiple frozen tissue arrays are designed for studying tissue specific cellular localization of genes or proteins of interest in different rabbit tissues. Up to 30 different frozen tissues from the same male or female rabbit are mounted on positively charged glass slides. Total RNA and cDNA from those tissues may also be available upon request.
Catalog No. 50-180-894