Learn More
Biochain Institute Inc Frozen Tissue Array - Human Tumor and Normal Tissue, Multi-tissue I, 1 slide/PK
SDP
Supplier:  Biochain Institute Inc T62357001
Encompass_Preferred
BioChain's tissue products are based on the sample repository network established following the IRB-approved ethical standard and procedures. The tissues are immediately put in liquid nitrogen after being excised, and then identified by a board-licensed pathologist. Patient demographic and disease related history data are available. BioChain's human tumor and normal multiple frozen tissue arrays are designed for studying tumor tissue specific cellular localization of genes or proteins of interest in different human tissues. 14 tumors and 14 correspondant normal tissues are mounted on a positively charged glass slide.
Catalog No. 50-180-885