Learn More
Biochain Institute Inc Frozen Tissue Array - Human Adult Normal Tissue, Multi-tissue I, 5 slides/PK
SDP
Catalog No. 50180875
Encompass_Preferred
BioChain's tissue products are based on the sample repository network established following the IRB-approved ethical standard and procedures. The tissues are immediately put in liquid nitrogen after being excised, and then identified by a board-licensed pathologist. Patient demographic and disease related history data is available. BioChain's Frozen Tissue Panels are manufactured using high-quality tissues.BioChain's Frozen Tissue Arrays are manufactured using high-quality tissues. Up to 45 different tissues are mounted on a modified glass slide after preparation with proprietary techniques. Each section is approximately 5 um thick. H.E. staining and immunostaining using beta-actin antibody ensure quality. Frozen tissue array is ideal for rapid cellular localization of RNA and protein expression.
Catalog No. 50-180-875 Supplier Biochain Institute Inc Supplier No. T62347005
BioChain's tissue products are based on the sample repository network established following the IRB-approved ethical standard and procedures. The tissues are immediately put in liquid nitrogen after being excised, and then identified by a board-licensed pathologist. Patient demographic and disease related history data is available. BioChain's Frozen Tissue Panels are manufactured using high-quality tissues.BioChain's Frozen Tissue Arrays are manufactured using high-quality tissues. Up to 45 different tissues are mounted on a modified glass slide after preparation with proprietary techniques. Each section is approximately 5 um thick. H.E. staining and immunostaining using beta-actin antibody ensure quality. Frozen tissue array is ideal for rapid cellular localization of RNA and protein expression.