Learn More
Biochain Institute Inc Frozen Tissue Array - Dog Male Adult Normal Tissue, Multi-tissue I, 1 slide/PK
SDP
Catalog No. 50180864
Encompass_Preferred
BioChain's dog normal multiple frozen tissue arrays are designed for studying tissue specific cellular localization of genes or proteins of interest in different dog tissues. Up to 30 different frozen tissues from the same male or female dog are mounted on positively charged glass slides. Total RNA and cDNA from those tissues may also be available upon request.
Catalog No. 50-180-864 Supplier Biochain Institute Inc Supplier No. T6734700G011
BioChain's dog normal multiple frozen tissue arrays are designed for studying tissue specific cellular localization of genes or proteins of interest in different dog tissues. Up to 30 different frozen tissues from the same male or female dog are mounted on positively charged glass slides. Total RNA and cDNA from those tissues may also be available upon request.