Learn More
Biochain Institute Inc FDA Standard Frozen Tissue Array - Mouse Normal, 4 slides/PK
SDP
Catalog No. 50180871
Encompass_Preferred
BioChain provides FDA Standard Frozen Tissue Array which is designed in conformance with FDA guideline and meeting the requirements for IHC (Immunohistochemistry) and IVD (In vitro Diagnostic Devices) certification. This tissue array has two slides which contain 30 different normal tissue types and 3 donors per tissue type.
Catalog No. 50-180-871 Supplier Biochain Institute Inc Supplier No. T63347012
BioChain provides FDA Standard Frozen Tissue Array which is designed in conformance with FDA guideline and meeting the requirements for IHC (Immunohistochemistry) and IVD (In vitro Diagnostic Devices) certification. This tissue array has two slides which contain 30 different normal tissue types and 3 donors per tissue type.