Learn More
Biochain Institute Inc FDA Standard Frozen Tissue Array - Human Adult Normal, 2 slides/PK
SDP
Catalog No. 50180868
Encompass_Preferred
This FDA Standard Frozen Tissue Array is designed according to published Guidance for Submission of Immunohistochemistry Applications to the FDA. Each FDA Standard Frozen Tissue Array package has either two or four slides which contain 30 different human normal tissue types and 3 donors per tissue type. This product is intended for Research Use Only.
Catalog No. 50-180-868 Supplier Biochain Institute Inc Supplier No. T62347011
This FDA Standard Frozen Tissue Array is designed according to published Guidance for Submission of Immunohistochemistry Applications to the FDA. Each FDA Standard Frozen Tissue Array package has either two or four slides which contain 30 different human normal tissue types and 3 donors per tissue type. This product is intended for Research Use Only.