Learn More
Biochain Institute Inc Frozen Tissue Section - Human Adult Normal: Brain: Hippocampus, 5 slides/PK
SDP
Supplier:  Biochain Institute Inc T1234052
Encompass_Preferred
BioChain's tissue products are based on the sample repository network established following the IRB-approved ethical standard and procedures. BioChain's Frozen Tissue Slides are ideal for rapidly identifying cellular localization of RNA or protein. A single human adult normal tissue with 5-10 um thickness is mounted on a positively charged glass slide. The slides are fixed and dehydrated with acetone for consistent results with in situ hybridization and immunohistochemistry. More organs may be available upon request.
Catalog No. 50-180-936