Learn More
Biochain Institute Inc Frozen Tissue Section Panel - Mouse Normal Tissue, Multi-tissue II, 5 slides/PK
SDP
Catalog No. 50181331
Encompass_Preferred
BioChain's Balb/C mouse normal multiple frozen tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different mouse tissues. 8 different frozen tissues are mounted on positively charged glass slides. Total RNA Northern Blots are available in the identical format, and cDNA from those tissues may also be available for PCR detection.
Catalog No. 50-181-331 Supplier Biochain Institute Inc Supplier No. T6334608
BioChain's Balb/C mouse normal multiple frozen tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different mouse tissues. 8 different frozen tissues are mounted on positively charged glass slides. Total RNA Northern Blots are available in the identical format, and cDNA from those tissues may also be available for PCR detection.