Learn More
Biochain Institute Inc Frozen Tissue Section Panel - Human Disease Tissue, Alzheimer's Disease, Multi-tissue I, 7 different tissues, 5 slides/PK
SDP
Catalog No. 50181321
Encompass_Preferred
BioChain's neurological diseased frozen tissue panels are designed for studying Alzheimer, Parkinson's Disease, Dementia, Multiple Sclerosis, Progressive Supranuclear Palsy (PSP), and Depression diseases. 6-8 same diseased frozen tissues with 5-10 um thickness are mounted on a positively charged glass slide. The slides are fixed and dehydrated with acetone for consistent results with in situ hybridization and immunohistochemistry.
Catalog No. 50-181-321 Supplier Biochain Institute Inc Supplier No. T6236444ALZ
Add to Cart
Add to Cart
BioChain's neurological diseased frozen tissue panels are designed for studying Alzheimer, Parkinson's Disease, Dementia, Multiple Sclerosis, Progressive Supranuclear Palsy (PSP), and Depression diseases. 6-8 same diseased frozen tissues with 5-10 um thickness are mounted on a positively charged glass slide. The slides are fixed and dehydrated with acetone for consistent results with in situ hybridization and immunohistochemistry.