Learn More
Biochain Institute Inc Frozen Tissue Section Panel - Rat Normal Tissue, Multi-tissue I, 5 slides/PK
SDP
Catalog No. 50181333
Encompass_Preferred
BioChain's Sprague Dawley or Wistar rat normal multiple frozen tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different rat tissues. 8 different frozen tissues are mounted on positively charged glass slides. Total RNA Northern Blots are available in the identical format, and cDNA from those tissues may also be available for PCR detection.
Catalog No. 50-181-333 Supplier Biochain Institute Inc Supplier No. T6434448
BioChain's Sprague Dawley or Wistar rat normal multiple frozen tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different rat tissues. 8 different frozen tissues are mounted on positively charged glass slides. Total RNA Northern Blots are available in the identical format, and cDNA from those tissues may also be available for PCR detection.