Learn More
Biochain Institute Inc Frozen Tissue Section Panel - Monkey (Rhesus) Normal Tissue, Multi-tissue III, 5 slides/PK
SDP
Supplier:  Biochain Institute Inc T6534423
Encompass_Preferred
BioChain's Rhesus monkey normal multiple frozen tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different monkey tissues. 8 different frozen tissues are mounted on positively charged glass slides. Total RNA Northern Blots are available in the identical format, and cDNA from those tissues may also be available for PCR detection.
Catalog No. 50-181-327