Learn More
Biochain Institute Inc Frozen Tissue Section Panel - Human Adult Normal Tissue, Multi-tissue I, 5 slides/PK
SDP
Catalog No. 50181318
Encompass_Preferred
BioChain's tissue products are based on the sample repository network established following the IRB-approved ethical standard and procedures. The tissues are immediately put in liquid nitrogen after being excised, and then identified by a board-licensed pathologist. Patient demographic and disease related history data are available. BioChain's Frozen Tissue Panels are manufactured using high-quality tissues. 5 to 8 different tissues are mounted on a modified glass slide after preparation with proprietary techniques. Each section is approximately 5 um thick. H.E. staining, immunostaining, and in situ hybridization with proliferating cellular nuclear antigen (PCNA) antibody and P53 gene probe ensure the quality of tissue panels. It is an ideal product for rapid cellular localization of RNA and protein expression.
Catalog No. 50-181-318 Supplier Biochain Institute Inc Supplier No. T6234431
BioChain's tissue products are based on the sample repository network established following the IRB-approved ethical standard and procedures. The tissues are immediately put in liquid nitrogen after being excised, and then identified by a board-licensed pathologist. Patient demographic and disease related history data are available. BioChain's Frozen Tissue Panels are manufactured using high-quality tissues. 5 to 8 different tissues are mounted on a modified glass slide after preparation with proprietary techniques. Each section is approximately 5 um thick. H.E. staining, immunostaining, and in situ hybridization with proliferating cellular nuclear antigen (PCNA) antibody and P53 gene probe ensure the quality of tissue panels. It is an ideal product for rapid cellular localization of RNA and protein expression.