Learn More
Biochain Institute Inc Frozen Tissue Section Panel - Monkey (Cynomolgus) Normal Tissue, Multi-tissue I, 5 slides/PK
SDP
Catalog No. 50181326
Encompass_Preferred
BioChain's Cynomolgus monkey normal multiple frozen tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different monkey tissues. 8 different frozen tissues are mounted on positively charged glass slides. Total RNA Northern Blots are available in the identical format, and cDNA from those tissues may also be available for PCR detection.
Catalog No. 50-181-326 Supplier Biochain Institute Inc Supplier No. T6534448CY
Add to Cart
Add to Cart
BioChain's Cynomolgus monkey normal multiple frozen tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different monkey tissues. 8 different frozen tissues are mounted on positively charged glass slides. Total RNA Northern Blots are available in the identical format, and cDNA from those tissues may also be available for PCR detection.