Learn More
Biochain Institute Inc Frozen Tissue Section Panel - Human Disease Tissue, Alzheimer's Disease, Multi-tissue II, 7 different tissues, 5 slides/PK
SDP
Supplier:  Biochain Institute Inc T6236445ALZ
Encompass_Preferred
BioChain's neurological diseased frozen tissue panels are designed for studying Alzheimer, Parkinson's Disease, Dementia, Multiple Sclerosis, Progressive Supranuclear Palsy (PSP), and Depression diseases. 6-8 same diseased frozen tissues with 5-10 um thickness are mounted on a positively charged glass slide. The slides are fixed and dehydrated with acetone for consistent results with in situ hybridization and immunohistochemistry.
Catalog No. 50-181-322