Learn More
MSC Cooper Bussmann FRS-R-110 300 VDC, 600 VAC, 110 Amp, Time Delay General Purpose Fuse Bolt-on Mount, 9-5/8" Overall Length, 20 at DC, 200 (RMS) kA Rating, 1-13/16" Diam
Catalog No. 501305039
Encompass
Cooper Bussmann FRS-R-110 300 VDC, 600 VAC, 110 Amp, Time Delay General Purpose Fuse Bolt-on Mount, 9-5/8" Overall Length, 20 at DC, 200 (RMS) kA Rating, 1-13/16" Diam
Catalog No. 50-130-5039 Supplier MSC Supplier No. 75908558
Cooper Bussmann FRS-R-110 300 VDC, 600 VAC, 110 Amp, Time Delay General Purpose Fuse Bolt-on Mount, 9-5/8" Overall Length, 20 at DC, 200 (RMS) kA Rating, 1-13/16" Diam