Learn More
MSC Cooper Bussmann FRS-R-2-1/4 300 VDC, 600 VAC, 2.25 Amp, Time Delay General Purpose Fuse Fuse Holder Mount, 127mm Overall Length, 20 at DC, 200 (RMS) kA Rating, 13/16" Diam
Catalog No. 501305050
Encompass
Cooper Bussmann FRS-R-2-1/4 300 VDC, 600 VAC, 2.25 Amp, Time Delay General Purpose Fuse Fuse Holder Mount, 127mm Overall Length, 20 at DC, 200 (RMS) kA Rating, 13/16" Diam
Catalog No. 50-130-5050 Supplier MSC Supplier No. 75908657
Cooper Bussmann FRS-R-2-1/4 300 VDC, 600 VAC, 2.25 Amp, Time Delay General Purpose Fuse Fuse Holder Mount, 127mm Overall Length, 20 at DC, 200 (RMS) kA Rating, 13/16" Diam