Learn More
Cayman Chemical Peiminine, Purity: ≥98%, CAS: 18059-10-4, 10 mg
SDP
Catalog No. 502990521
Encompass_Preferred
An alkaloid with diverse biological activities; increases LC3B-II protein levels and induces cell death in HCT116 colorectal carcinoma cells in a concentration-dependent manner; reduces tumor growth in an HCT116 mouse xenograft model at 3 mg/kg every other day; has antitussive effects in a mouse cough model induced by ammonia liquor at3 mg/kg; reduces dermal and epidermal thickness, inhibits dermal infiltration of eosinophils and mast cells, and decreases serum levels of IgE, IL-4, and TNF-α in a mouse model of DNCB-induced atopic dermatitis at 5 mg/kg
Catalog No. 50-299-0521 Supplier Cayman Chemical Supplier No. 2961810 MG
An alkaloid with diverse biological activities; increases LC3B-II protein levels and induces cell death in HCT116 colorectal carcinoma cells in a concentration-dependent manner; reduces tumor growth in an HCT116 mouse xenograft model at 3 mg/kg every other day; has antitussive effects in a mouse cough model induced by ammonia liquor at3 mg/kg; reduces dermal and epidermal thickness, inhibits dermal infiltration of eosinophils and mast cells, and decreases serum levels of IgE, IL-4, and TNF-α in a mouse model of DNCB-induced atopic dermatitis at 5 mg/kg