Learn More
Grainger FORTUNA Reusable Glass Syringe, 10-19G
Catalog No. NC2584234
Encompass_Preferred
The Fortuna Reusable Glass Syringe is a 10mL syringe made of heat-resistant borosilicate glass with a Luer lock tip. It has clear barrel markings and an ergonomic flange handle design. This reusable syringe can withstand temperatures up to 500°F/260°C and is autoclavable for sterilization. It has a stainless steel smooth plunger with a PTFE gasket tip. The syringe is latex-free. This product can be used for food applications as well as scientific and industrial uses. It meets ISO 7886-1 standards.
Catalog No. NC2584234 Supplier Grainger Supplier No. 19G352
The Fortuna Reusable Glass Syringe is a 10mL syringe made of heat-resistant borosilicate glass with a Luer lock tip. It has clear barrel markings and an ergonomic flange handle design. This reusable syringe can withstand temperatures up to 500°F/260°C and is autoclavable for sterilization. It has a stainless steel smooth plunger with a PTFE gasket tip. The syringe is latex-free. This product can be used for food applications as well as scientific and industrial uses. It meets ISO 7886-1 standards.