Learn More
Medchemexpress LLC Fructo-oligosaccharide DP8/GF7 | 62512-21-4 | 5 MG
SDP
Catalog No. 5000418961
Encompass_Preferred
Fructo-oligosaccharide DP8/GF7 is a fructooligosaccharide (FOS) with a degree of polymerization (DP) of 8. It consists of 7 fructose units connected by (2→1)-β-glycosidic bonds and includes a single D-glucosyl unit at its non-reducing end.
  • Belongs to fructooligosaccharides (FOS)
  • Has a degree of polymerization (DP) of 8
  • Composed of 7 fructose units linked by (2→1)-β-glycosidic bonds
  • Contains a single D-glucosyl unit at the non-reducing end
Catalog No. 50-004-18961 Supplier Medchemexpress LLC Supplier No. HYN68365MG
Fructo-oligosaccharide DP8/GF7 is a fructooligosaccharide (FOS) with a degree of polymerization (DP) of 8. It consists of 7 fructose units connected by (2→1)-β-glycosidic bonds and includes a single D-glucosyl unit at its non-reducing end.
  • Belongs to fructooligosaccharides (FOS)
  • Has a degree of polymerization (DP) of 8
  • Composed of 7 fructose units linked by (2→1)-β-glycosidic bonds
  • Contains a single D-glucosyl unit at the non-reducing end