Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fructo-oligosaccharide DP9/GF8 | 143625-74-5 | 5 MG
SDP

Catalog No. 5000419742
Encompass_Preferred

Fructo-oligosaccharide DP9/GF8 is a fructooligosaccharide (FOS) characterized by a degree of polymerization (DP) of 9. These FOS molecules consist of 8 fructose units connected by (2→1)-β-glycosidic bonds and feature a single D-glucosyl unit at the non-reducing end. This product is designed strictly for research purposes.

  • Purity: 99.49%
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Saccharides, polysaccharides
  • Initial source: Plants, Gramineae, Saccharum officinarum

Catalog No. 50-004-19742 Supplier Medchemexpress LLC Supplier No. HYN68355MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fructo-oligosaccharide DP9/GF8 is a fructooligosaccharide (FOS) characterized by a degree of polymerization (DP) of 9. These FOS molecules consist of 8 fructose units connected by (2→1)-β-glycosidic bonds and feature a single D-glucosyl unit at the non-reducing end. This product is designed strictly for research purposes.

  • Purity: 99.49%
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Saccharides, polysaccharides
  • Initial source: Plants, Gramineae, Saccharum officinarum

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.