Learn More
FUJIFILM Biosciences Fructooligosaccharides Set
Catalog No. 5000450325
Encompass_Preferred
This set consists of 250 mg vial each of 1-Kestose Nystose and 1F-Fructofuranosylnystose It is applicable to analyze the fructooligosaccharide components contained in foods and beverages For HPLC
Catalog No. 50-004-50325 Supplier FUJIFILM Biosciences Supplier No. 29573401
This set consists of 250 mg vial each of 1-Kestose Nystose and 1F-Fructofuranosylnystose It is applicable to analyze the fructooligosaccharide components contained in foods and beverages For HPLC