Learn More
Randox Fructosamine Chemical Reagent, Liquid, ready-to-use, 5 x 25 mL R1 Volume, 5 x 6.3 mL R2 Volume, Measuring Range 8.12 - 1803 μmol/l, Storage Temperature 2-8°C, Sample Type Human serum or plasma
SDP
Catalog No. NC2160601
Encompass_Preferred
Fructosamine Chemical Reagent, Liquid, ready-to-use, 5 x 25 mL R1 Volume, 5 x 6.3 mL R2 Volume, Measuring Range 8.12 - 1803 μmol/l, Storage Temperature 2-8°C, Sample Type Human serum or plasma
This item is not returnable. View return policy
Catalog No. NC2160601 Supplier Randox Supplier No. FR3133
 Restricted Item
Add to Cart
Add to Cart
Fructosamine Chemical Reagent, Liquid, ready-to-use, 5 x 25 mL R1 Volume, 5 x 6.3 mL R2 Volume, Measuring Range 8.12 - 1803 μmol/l, Storage Temperature 2-8°C, Sample Type Human serum or plasma