Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fructosamine hydrochloride | 39002-30-7 | 96.9% | 215.63 g/mol | C6H14ClNO5 | 50 MG
SDP

Catalog No. 5000281343
Encompass_Preferred

Fructosamine hydrochloride is the hydrochloride salt of an amino monosaccharide provided as a research reagent. It is described as a metabolic intermediate and structural component relevant to cell membrane and cartilage studies, and has been reported to cause site-specific DNA strand breaks in the presence of Cu2+.

  • Research-grade amino sugar hydrochloride reagent.
  • CAS number 39002-30-7.
  • Molecular formula C6H14ClNO5; molecular weight 215.63 g/mol.
  • Purity 96.9% as provided by the manufacturer.
  • Available in multiple sizes, including 50 mg packaging.
  • Useful for biochemical and cellular research involving metabolic intermediates and structural studies.
  • May induce DNA strand breaks in the presence of copper; follow appropriate safety procedures.

Catalog No. 50-002-81343 Supplier Medchemexpress LLC Supplier No. HYW41432150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fructosamine hydrochloride is the hydrochloride salt of an amino monosaccharide provided as a research reagent. It is described as a metabolic intermediate and structural component relevant to cell membrane and cartilage studies, and has been reported to cause site-specific DNA strand breaks in the presence of Cu2+.

  • Research-grade amino sugar hydrochloride reagent.
  • CAS number 39002-30-7.
  • Molecular formula C6H14ClNO5; molecular weight 215.63 g/mol.
  • Purity 96.9% as provided by the manufacturer.
  • Available in multiple sizes, including 50 mg packaging.
  • Useful for biochemical and cellular research involving metabolic intermediates and structural studies.
  • May induce DNA strand breaks in the presence of copper; follow appropriate safety procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.