Learn More
Medchemexpress LLC Fructosamine hydrochloride | 39002-30-7 | ≥97.0% | 215.63 g/mol | C6H14ClNO5 | 10 MG
SDP
Catalog No. 5000281126
Encompass_Preferred
Fructosamine hydrochloride is an amino sugar hydrochloride used as a research reagent in biochemical and cell biology studies. It serves as a metabolic intermediate and a component of cell membranes and cartilage, and has been reported to induce site-specific DNA strand breaks in the presence of copper ions. Supplied as a solid with specified purity and storage recommendations, it is intended for laboratory research use.
  • Metabolic intermediate useful for biochemical studies.
  • Component of cell membranes and cartilage-related research.
  • Reported to induce site-specific DNA strand breaks in the presence of copper ions (Cu2+).
  • Available in small milligram pack sizes for assay development.
  • High purity ≥97.0% suitable for research applications.
  • Solid, white to off-white appearance; easy to weigh and handle.
  • Store sealed, away from moisture; follow cold storage recommendations for solutions.
Catalog No. 50-002-81126 Supplier Medchemexpress LLC Supplier No. HYW41432110MG
Fructosamine hydrochloride is an amino sugar hydrochloride used as a research reagent in biochemical and cell biology studies. It serves as a metabolic intermediate and a component of cell membranes and cartilage, and has been reported to induce site-specific DNA strand breaks in the presence of copper ions. Supplied as a solid with specified purity and storage recommendations, it is intended for laboratory research use.
  • Metabolic intermediate useful for biochemical studies.
  • Component of cell membranes and cartilage-related research.
  • Reported to induce site-specific DNA strand breaks in the presence of copper ions (Cu2+).
  • Available in small milligram pack sizes for assay development.
  • High purity ≥97.0% suitable for research applications.
  • Solid, white to off-white appearance; easy to weigh and handle.
  • Store sealed, away from moisture; follow cold storage recommendations for solutions.