Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fructosamine hydrochloride | 39002-30-7 | >97.0% | 215.63 g/mol | C6H14ClNO5 | 25 MG
SDP

Catalog No. 5000281186
Encompass_Preferred

Fructosamine hydrochloride is the hydrochloride salt of D-isoglucosamine supplied as a white to off-white solid for biochemical research. It is used as a reagent and metabolic intermediate in studies of protein glycation, cell membrane biology, and related biochemical processes. Handle per standard laboratory safety practices.

  • CAS number: 39002-30-7.
  • Molecular formula: C6H14ClNO5.
  • Molecular weight: 215.63 g/mol.
  • Purity: ≥97.0%.
  • Appearance: white to off-white solid.
  • Storage: store sealed at 4°C; stock solutions -80°C (6 months) or -20°C (1 month).
  • Pack sizes: available in mg-scale packs including 25 mg.

Catalog No. 50-002-81186 Supplier Medchemexpress LLC Supplier No. HYW41432125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fructosamine hydrochloride is the hydrochloride salt of D-isoglucosamine supplied as a white to off-white solid for biochemical research. It is used as a reagent and metabolic intermediate in studies of protein glycation, cell membrane biology, and related biochemical processes. Handle per standard laboratory safety practices.

  • CAS number: 39002-30-7.
  • Molecular formula: C6H14ClNO5.
  • Molecular weight: 215.63 g/mol.
  • Purity: ≥97.0%.
  • Appearance: white to off-white solid.
  • Storage: store sealed at 4°C; stock solutions -80°C (6 months) or -20°C (1 month).
  • Pack sizes: available in mg-scale packs including 25 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.