Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fructosamine hydrochloride | 39002-30-7 | MFCD27981318 | ≥97.0% | 215.63 g/mol | C6H14ClNO5 | 5 MG
SDP

Catalog No. 5000281688
Encompass_Preferred

Fructosamine hydrochloride is the hydrochloride salt of an amino sugar used as a metabolic intermediate and as a component of cell membranes and cartilage. Supplied as a solid research reagent, it is employed in biochemical studies and has been reported to induce site-specific DNA strand breaks.

  • High purity suitable for research: ≥97.0%.
  • Solid form for easy storage and handling.
  • Molecular weight 215.63 g/mol.
  • CAS number 39002-30-7 for unambiguous identification.
  • Available in small quantities for laboratory experiments, such as 5 mg.

Catalog No. 50-002-81688 Supplier Medchemexpress LLC Supplier No. HYW4143215MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fructosamine hydrochloride is the hydrochloride salt of an amino sugar used as a metabolic intermediate and as a component of cell membranes and cartilage. Supplied as a solid research reagent, it is employed in biochemical studies and has been reported to induce site-specific DNA strand breaks.

  • High purity suitable for research: ≥97.0%.
  • Solid form for easy storage and handling.
  • Molecular weight 215.63 g/mol.
  • CAS number 39002-30-7 for unambiguous identification.
  • Available in small quantities for laboratory experiments, such as 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.