Learn More
Apexbio Technology LLC Fructose 7660-25-5 20mg
SDP
Catalog No. 502240562
Encompass_Preferred
Fructose is a monosaccharide commonly used as an energy and carbon source in biochemical studies Fructose participates in metabolic pathways such as glycolysis and gluconeogenesis where it is enzymatically converted into intermediates like fructose-1-phosphate before entering carbohydrate metabolism Fructose is frequently applied as a nutrient in microbial fermentation media and mammalian cell culture systems Based on these properties fructose is utilized in research fields related to carbohydrate metabolism insulin sensitivity obesity diabetes and metabolic syndrome
This item is not returnable. View return policy
Catalog No. 50-224-0562 Supplier Apexbio Technology LLC Supplier No. N150320
Fructose is a monosaccharide commonly used as an energy and carbon source in biochemical studies Fructose participates in metabolic pathways such as glycolysis and gluconeogenesis where it is enzymatically converted into intermediates like fructose-1-phosphate before entering carbohydrate metabolism Fructose is frequently applied as a nutrient in microbial fermentation media and mammalian cell culture systems Based on these properties fructose is utilized in research fields related to carbohydrate metabolism insulin sensitivity obesity diabetes and metabolic syndrome