Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fructose 6 phosphate kinase antibody (YA1715) | 100 UL
SDP

Catalog No. 5000441021
Encompass_Preferred

Fructose 6 phosphate kinase antibody (YA1715) is a rabbit-derived and non-conjugated IgG monoclonal antibody, targeting Fructose 6 phosphate kinase. Three phosphofructokinase isozymes exist in humans: muscle, liver and platelet. These isozymes function as subunits of the mammalian tetramer phosphofructokinase, which catalyzes the phosphorylation of fructose-6-phosphate to fructose-1,6-bisphosphate. Tetramer composition varies depending on tissue type.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Conjugation: Non-conjugated
  • Application: WB (1:500-1:1000), ICC/IF (1:50-1:200)
  • Formulation: Supplied in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Purity: Affinity purified
  • Appearance: Liquid
  • Storage & stability: Stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles.

Catalog No. 50-004-41021 Supplier Medchemexpress LLC Supplier No. HYP81970100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fructose 6 phosphate kinase antibody (YA1715) is a rabbit-derived and non-conjugated IgG monoclonal antibody, targeting Fructose 6 phosphate kinase. Three phosphofructokinase isozymes exist in humans: muscle, liver and platelet. These isozymes function as subunits of the mammalian tetramer phosphofructokinase, which catalyzes the phosphorylation of fructose-6-phosphate to fructose-1,6-bisphosphate. Tetramer composition varies depending on tissue type.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Conjugation: Non-conjugated
  • Application: WB (1:500-1:1000), ICC/IF (1:50-1:200)
  • Formulation: Supplied in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Purity: Affinity purified
  • Appearance: Liquid
  • Storage & stability: Stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.