Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fructose | 7660-25-5 | 180.16 | 1 ML
SDP

Catalog No. 5000389967
Encompass_Preferred

Fructose is a simple ketonic monosaccharide commonly found in plants. It is for research use only. This product is supplied as a 10 mM solution in DMSO.

  • Simple ketonic monosaccharide
  • Purity of 99.92%
  • Appears as a white to off-white solid
  • Storage in solvent at -80°C for 2 years or -20°C for 1 year
  • Soluble in water and DMSO

Catalog No. 50-003-89967 Supplier Medchemexpress LLC Supplier No. HYN039510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fructose is a simple ketonic monosaccharide commonly found in plants. It is for research use only. This product is supplied as a 10 mM solution in DMSO.

  • Simple ketonic monosaccharide
  • Purity of 99.92%
  • Appears as a white to off-white solid
  • Storage in solvent at -80°C for 2 years or -20°C for 1 year
  • Soluble in water and DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.