Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fructose | 7660-25-5 | 180.16 | 25 MG
SDP

Catalog No. 5000389962
Encompass_Preferred

Fructose is a simple ketonic monosaccharide found in many plants, where it is often bonded to glucose to form the disaccharide sucrose.

  • Purity: 99.92%
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Plants other families
  • For research use only.

Catalog No. 50-003-89962 Supplier Medchemexpress LLC Supplier No. 25MGHYN0395R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fructose is a simple ketonic monosaccharide found in many plants, where it is often bonded to glucose to form the disaccharide sucrose.

  • Purity: 99.92%
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Plants other families
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.