Learn More
Medchemexpress LLC Fructose | 7660-25-5 | MFCD00065409 | 99.2% | 180.16 g/mol | C6H12O6 | 10 MG

Supplier: Medchemexpress LLC HYN0395R10MG
Fructose (standard) is an analytical reference material of the monosaccharide fructose supplied as a small packaged powder for research and analytical applications, including chromatography and quantitative assays. The material has a specified assay and recommended storage for laboratory use.
- Analytical reference standard for fructose.
- Packaged as a 10 mg powder.
- High purity assay (~99.2% reported on COA).
- Molecular weight 180.16 g/mol.
- Recommended storage at 4°C, protected from light and moisture.
- Intended for chromatography and quantitative analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.