Learn More
Creative Biomart Fructose Assay Kit is a quantitative colorimetric fructose determination at 565 nm.
Catalog No. NC3451524
Encompass
Direct Assays: fructose in biological samples (e.g. serum, plasma, urine, saliva, milk, culture medium), food, juice, beverage and other agricultural products.Drug Discovery/Pharmacology: effects of drugs on fructose metabolism.Storage : Store all components at -20°C. Shelf life of six months after receipt.
Catalog No. NC3451524 Supplier Creative Biomart Supplier No. KIT0344
Direct Assays: fructose in biological samples (e.g. serum, plasma, urine, saliva, milk, culture medium), food, juice, beverage and other agricultural products.Drug Discovery/Pharmacology: effects of drugs on fructose metabolism.Storage : Store all components at -20°C. Shelf life of six months after receipt.