Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fructose-bisphosphate aldolase A/ALDOA protein, human (His) | 96.0% | 10UG
SDP

Catalog No. 5000177380
Encompass_Preferred

Fructose-bisphosphate aldolase A (ALDOA) is a recombinant human glycolytic enzyme (364 amino acids) that catalyzes the reversible cleavage of β-D-fructose 1,6-bisphosphate into two triose phosphates. The protein is expressed in E. coli, supplied with an N-terminal 6xHis tag, and intended for research use.

  • Recombinant human ALDOA (364 aa) with N-terminal 6xHis tag.
  • Expressed in Escherichia coli and purified to high purity (COA reports 96% by SDS-PAGE).
  • Molecular weight approximately 45.3 kDa.
  • Suitable for enzyme assays, structural biology, and antibody production.
  • Available in small and larger sizes to support assay development and scaling.
  • Provided for research use only.

Catalog No. 50-001-77380 Supplier Medchemexpress LLC Supplier No. HYP70026510UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fructose-bisphosphate aldolase A (ALDOA) is a recombinant human glycolytic enzyme (364 amino acids) that catalyzes the reversible cleavage of β-D-fructose 1,6-bisphosphate into two triose phosphates. The protein is expressed in E. coli, supplied with an N-terminal 6xHis tag, and intended for research use.

  • Recombinant human ALDOA (364 aa) with N-terminal 6xHis tag.
  • Expressed in Escherichia coli and purified to high purity (COA reports 96% by SDS-PAGE).
  • Molecular weight approximately 45.3 kDa.
  • Suitable for enzyme assays, structural biology, and antibody production.
  • Available in small and larger sizes to support assay development and scaling.
  • Provided for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.