Learn More
Medchemexpress LLC Fructose-bisphosphate aldolase A/ALDOA protein, human (His) | >95.0% | 500UG
SDP
Catalog No. 5000177461
Encompass_Preferred
Recombinant human fructose-bisphosphate aldolase A (ALDOA) is supplied as an N-terminal His-tagged protein in lyophilized form for biochemical and cell biology research. The protein is expressed in Escherichia coli, provided with high purity and low endotoxin, and is suitable for enzymatic assays, structural studies, and antibody generation.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Lyophilized powder formulation for extended shelf life.
  • N-terminal 6xHis tag enables affinity purification and detection.
  • Low endotoxin level (<1 EU/μg) suitable for many in vitro applications.
  • Approximate molecular weight 45.3 kDa; full length 363 amino acids.
  • Recommended reconstitution at ≥100 μg/mL and storage in aliquots at -20°C or -80°C.
Catalog No. 50-001-77461 Supplier Medchemexpress LLC Supplier No. HYP700265500UG
Recombinant human fructose-bisphosphate aldolase A (ALDOA) is supplied as an N-terminal His-tagged protein in lyophilized form for biochemical and cell biology research. The protein is expressed in Escherichia coli, provided with high purity and low endotoxin, and is suitable for enzymatic assays, structural studies, and antibody generation.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Lyophilized powder formulation for extended shelf life.
  • N-terminal 6xHis tag enables affinity purification and detection.
  • Low endotoxin level (<1 EU/μg) suitable for many in vitro applications.
  • Approximate molecular weight 45.3 kDa; full length 363 amino acids.
  • Recommended reconstitution at ≥100 μg/mL and storage in aliquots at -20°C or -80°C.