Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fructose-L-tryptophan | 25020-15-9 | MF183634 | >95.0% | 366.37 g/mol | C17H22N2O7 | 1 MG
SDP

Catalog No. 5000284445
Encompass_Preferred

Fructose-L-tryptophan (Trp-Fru) is an early Maillard reaction product formed from L-tryptophan and simple sugars. It is supplied as a research-grade biochemical reagent for use in studies of protein glycation, Maillard chemistry, and related biochemical assays.

  • Early Maillard reaction product formed from tryptophan and glucose
  • Molecular formula C17H22N2O7; molecular weight 366.37 g/mol
  • CAS number 25020-15-9 for unambiguous identification
  • Available in small research quantities (1 mg, 5 mg)
  • Reported purity >95.0% (HPLC) for mixture of diastereomers
  • Store according to the certificate of analysis; follow handling documentation

Catalog No. 50-002-84445 Supplier Medchemexpress LLC Supplier No. HYW5858871MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fructose-L-tryptophan (Trp-Fru) is an early Maillard reaction product formed from L-tryptophan and simple sugars. It is supplied as a research-grade biochemical reagent for use in studies of protein glycation, Maillard chemistry, and related biochemical assays.

  • Early Maillard reaction product formed from tryptophan and glucose
  • Molecular formula C17H22N2O7; molecular weight 366.37 g/mol
  • CAS number 25020-15-9 for unambiguous identification
  • Available in small research quantities (1 mg, 5 mg)
  • Reported purity >95.0% (HPLC) for mixture of diastereomers
  • Store according to the certificate of analysis; follow handling documentation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.