Learn More
Medchemexpress LLC Fructose-phenylalanine | 31105-03-0 | MFCD30719642 | 93.0% | 327.33 | C15H21NO7 | 5 MG
SDP
Catalog No. 5000284915
Encompass_Preferred
Fructose-phenylalanine is a small-molecule research reagent reported as a non-competitive angiotensin-converting enzyme (ACE) inhibitor (IC50 = 0.19 mM) and shown to have antioxidant activity. It is supplied as a purified solid for laboratory use and is typically applied in biochemical assays, mechanistic studies, and analytical workflows.
  • Non-competitive ACE inhibitor (IC50 = 0.19 mM).
  • Exhibits antioxidant activity.
  • Suitable for biochemical assays and mechanistic studies.
  • Provided as a purified solid for laboratory use.
  • Molecular weight 327.33 g/mol; formula C15H21NO7.
  • Available in small package sizes for research-scale experiments.
Catalog No. 50-002-84915 Supplier Medchemexpress LLC Supplier No. HYW5859345MG
Fructose-phenylalanine is a small-molecule research reagent reported as a non-competitive angiotensin-converting enzyme (ACE) inhibitor (IC50 = 0.19 mM) and shown to have antioxidant activity. It is supplied as a purified solid for laboratory use and is typically applied in biochemical assays, mechanistic studies, and analytical workflows.
  • Non-competitive ACE inhibitor (IC50 = 0.19 mM).
  • Exhibits antioxidant activity.
  • Suitable for biochemical assays and mechanistic studies.
  • Provided as a purified solid for laboratory use.
  • Molecular weight 327.33 g/mol; formula C15H21NO7.
  • Available in small package sizes for research-scale experiments.