Learn More
Apexbio Technology LLC Fruquintinib(HMPL-013), 10mg. Cas: 1194506-26-7 MFCD: MFCD28502149. Potent and selective inhibitor of VEGFR 1, 2, 3
SDP
Supplier:  Apexbio Technology LLC B586410
Encompass_Preferred
IC50: 33 nmol/L, 35 nmol/L and 0.5 nmol/L for VEGFR1, 2, 3VEGF/VEGFR signal has been proven to be an important target for development of novel cancer therapies. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5369