Learn More
Apexbio Technology LLC Fruquintinib(HMPL-013) 1194506-26-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063641
Encompass_Preferred
Fruquintinib (HMPL-013 CAS 1194506-26-7) is a small molecule inhibitor targeting VEGFR family receptor tyrosine kinases (VEGFR1 VEGFR2 and VEGFR3) It demonstrates potent inhibitory activity with reported IC50 values of approximately 33 nM (VEGFR1) 35 nM (VEGFR2) and 0 5 nM (VEGFR3) Kinase selectivity screening indicates limited off-target effects against other kinases such as RET FGFR-1 and c-kit In preclinical tumor xenograft studies (e g gastric cancer BGC-823 model) significant tumor growth suppression ( 80%) was achieved at sustained therapeutic exposure Fruquintinib is currently under clinical investigation for cancer treatment based on its selective VEGFR pathway inhibition
Catalog No. 50-000-63641 Supplier Apexbio Technology LLC Supplier No. B58645.1
Fruquintinib (HMPL-013 CAS 1194506-26-7) is a small molecule inhibitor targeting VEGFR family receptor tyrosine kinases (VEGFR1 VEGFR2 and VEGFR3) It demonstrates potent inhibitory activity with reported IC50 values of approximately 33 nM (VEGFR1) 35 nM (VEGFR2) and 0 5 nM (VEGFR3) Kinase selectivity screening indicates limited off-target effects against other kinases such as RET FGFR-1 and c-kit In preclinical tumor xenograft studies (e g gastric cancer BGC-823 model) significant tumor growth suppression ( 80%) was achieved at sustained therapeutic exposure Fruquintinib is currently under clinical investigation for cancer treatment based on its selective VEGFR pathway inhibition