Learn More
Grainger Fry Pan Handle Rubber
Catalog No. 502763759
Encompass_Preferred
The Crestware Fry Pan Handle is a replacement handle for 10-inch and 12-inch fry pans. It is made of stainless steel with a gray powder coat finish and measures 11 inches long. The handle attaches with two stainless steel rivets and has a hole for hanging storage. It can withstand oven temperatures up to 500°F. This sturdy handle provides a secure grip and even heating. It's a high quality replacement handle to fix or upgrade old fry pans.
Catalog No. 50-276-3759 Supplier Grainger Supplier No. 16W520
The Crestware Fry Pan Handle is a replacement handle for 10-inch and 12-inch fry pans. It is made of stainless steel with a gray powder coat finish and measures 11 inches long. The handle attaches with two stainless steel rivets and has a hole for hanging storage. It can withstand oven temperatures up to 500°F. This sturdy handle provides a secure grip and even heating. It's a high quality replacement handle to fix or upgrade old fry pans.