Learn More
Grainger Fryer Basket 4.75 x 8.75 x 5.25 In PK12
Catalog No. 502753564
Encompass_Preferred
The Grainger Fryer Basket is made of stainless steel with a wire mesh design. It has dimensions of 4-3/4" x 8-3/4" x 5-1/4" and is designed to fit most commercial deep fryers. This fryer basket allows efficient frying by maximizing oil circulation. It features a sturdy handle for safe insertion and removal from hot oil. The wire mesh allows for quick, even cooking and drainage of oil. This fryer basket is NSF certified.
Catalog No. 50-275-3564 Supplier Grainger Supplier No. 14D424
The Grainger Fryer Basket is made of stainless steel with a wire mesh design. It has dimensions of 4-3/4" x 8-3/4" x 5-1/4" and is designed to fit most commercial deep fryers. This fryer basket allows efficient frying by maximizing oil circulation. It features a sturdy handle for safe insertion and removal from hot oil. The wire mesh allows for quick, even cooking and drainage of oil. This fryer basket is NSF certified.