Learn More
Cole-Parmer Argos Technologies PolarSafe Vertical/Chest Freezer Rack for Standard 3.75" High Boxes, 5 box capacity
Catalog No. 501960814 Shop All Cole-Parmer Products
Encompass_Preferred
These racks to store 3.75-inch high polycarbonate boxes come with locking rods to securely store boxes in place. The racks have sturdy dual stainless steel handles for easy handling, and made of corrosion-resistant stainless steel.
Catalog No. 50-196-0814 Supplier Cole-Parmer Supplier No. 0440446
These racks to store 3.75-inch high polycarbonate boxes come with locking rods to securely store boxes in place. The racks have sturdy dual stainless steel handles for easy handling, and made of corrosion-resistant stainless steel.