Learn More
Cole-Parmer Argos Technologies PolarSafe Vertical/Chest Rack for Large 3.75" High Boxes, 5 box capacity
Catalog No. 501960818 Shop All Cole-Parmer Products
Encompass_Preferred
These racks store large boxes that measure 3-3/4" x 5-13/16" x 5-13/16" come with locking rods to securely keep them in place. The racks have sturdy dual stainless steel handles for easy handling, and made of corrosion-resistant stainless steel.
Catalog No. 50-196-0818 Supplier Cole-Parmer Supplier No. 0440474
Add to Cart
Add to Cart
These racks store large boxes that measure 3-3/4" x 5-13/16" x 5-13/16" come with locking rods to securely keep them in place. The racks have sturdy dual stainless steel handles for easy handling, and made of corrosion-resistant stainless steel.