Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for Microtube Boxes with 2 7/16" Max Height, 6 x 3 Array
Catalog No. 501960770 Shop All Cole-Parmer Products
Encompass_Preferred
These upright freezer racks come with a locking rod to securely hold boxes in place. Boxes can be accessed from either side of the rack, which has handles on both sides to accommodate freezers with left- or right-handed doors. They are crafted of corrosion-resist stainless steel for long-term use.
Catalog No. 50-196-0770 Supplier Cole-Parmer Supplier No. 0440202
These upright freezer racks come with a locking rod to securely hold boxes in place. Boxes can be accessed from either side of the rack, which has handles on both sides to accommodate freezers with left- or right-handed doors. They are crafted of corrosion-resist stainless steel for long-term use.