Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for Microtube Boxes with 2 7/16" Max Height, 6 x 4 Array
Catalog No. 501961833 Shop All Cole-Parmer Products
Encompass_Preferred
Freezer includes locking rods that securely hold boxes in place. Access boxes from either side of rack Handles on both sides accommodate freezers with left- or right-handed door Sturdy corrosion-resistant stainless steel construction for long-term use
Catalog No. 50-196-1833 Supplier Cole-Parmer Supplier No. 0440203
Freezer includes locking rods that securely hold boxes in place. Access boxes from either side of rack Handles on both sides accommodate freezers with left- or right-handed door Sturdy corrosion-resistant stainless steel construction for long-term use