Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for Microtube Boxes with 2 7/16" Max Height, 6 x 5 Array
Catalog No. 501960521 Shop All Cole-Parmer Products
Encompass_Preferred
These upright freezer racks with a locking rod are designed for storing microtube boxes. They are crafted of corrosion-resistent stainless steel for durablity.
Catalog No. 50-196-0521 Supplier Cole-Parmer Supplier No. 0440204
These upright freezer racks with a locking rod are designed for storing microtube boxes. They are crafted of corrosion-resistent stainless steel for durablity.