Learn More
Aviva Systems Biology Corp FRZB Antibody - N-terminal region (ARP54570_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54570_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FRZB. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human FRZB. Alternate names: FRE, FRITZ, FRP-3, FRZB-1, FRZB-PEN, FRZB1, FZRB, OS1, SFRP3, SRFP3, hFIZApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-941