Learn More
Aviva Systems Biology Corp FRZB Antibody - middle region (ARP54571_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54571_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FRZB. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human FRZB. Alternate names: FRE, FRITZ, FRP-3, FRZB-1, FRZB-PEN, FRZB1, FZRB, SFRP3, SRFP3, hFIZ, OS1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-942