Learn More
Promocell GmbH Cryo-SFM Plus, Freezing Medium, 125 ml, Defined, Animal Components Free, Protein Content Free, Methylcellulose, DMSO, Antioxidants, Primary human cells, Stem cells, Cell lines, Cell aggregates
Catalog No. NC3367467
Encompass
Cryo-SFM Plus, Freezing Medium, 125 ml, Defined, Animal Components Free, Protein Content Free, Methylcellulose, DMSO, Antioxidants, Primary human cells, Stem cells, Cell lines, Cell aggregates
Catalog No. NC3367467 Supplier Promocell GmbH Supplier No. C29922
Cryo-SFM Plus, Freezing Medium, 125 ml, Defined, Animal Components Free, Protein Content Free, Methylcellulose, DMSO, Antioxidants, Primary human cells, Stem cells, Cell lines, Cell aggregates